missing translation for 'onlineSavingsMsg'
Learn More

2-(4-Methoxyphenyl)acetyl chloride, TRC

Product Code. 30475418
Change view
Click to view available options
Quantity:
2.5 g
Unit Size:
2.5g
Product Code. Quantity unitSize
30475418 2.5 g 2.5g
1 options
This item is not returnable. View return policy

Product Code. 30475418

Brand: Toronto Research Chemicals TRCM9452432.5G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-(4-Methoxyphenyl)acetyl chloride
CAS 4693-91-8
Molecular Formula C9H9O2Cl
Quantity 2.5 g
InChI Formula InChI=1S/C9H9ClO2/c1-12-8-4-2-7(3-5-8)6-9(10)11/h2-5H,6H2,1H3
SMILES COC1=CC=C(C=C1)CC(=O)Cl
IUPAC Name 2-(4-Methoxyphenyl)acetyl Chloride
Molecular Weight (g/mol) 184.61
Formula Weight 184.03
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.