missing translation for 'onlineSavingsMsg'
Learn More

2-(4-Methylphenyl)ethanol, 99%

Product Code. 11459956
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11459956 25 g 25g
11439956 1 g 1g
11449956 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11459956 Supplier Thermo Scientific Alfa Aesar Supplier No. B20918.14
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 699-02-5 | C9H12O | 136.194 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 699-02-5
Molecular Formula C9H12O
Molecular Weight (g/mol) 136.194
MDL Number MFCD00002904
InChI Key DAVFJRVIVZOKKS-UHFFFAOYSA-N
Synonym 2-4-methylphenyl ethanol, 2-p-tolyl ethanol, 4-methylphenethyl alcohol, benzeneethanol, 4-methyl, 2-p-tolylethanol, p-methylphenethyl alcohol, 4-methylbenzeneethanol, 2-4-methylphenyl ethan-1-ol, p-methylphenylethyl alcohol, 2-p-methylphenyl ethanol
PubChem CID 95294
IUPAC Name 2-(4-methylphenyl)ethanol
SMILES CC1=CC=C(C=C1)CCO

Specifications

Density 0.98
Boiling Point 244°C to 245°C
Flash Point 107°C (225°F)
Refractive Index 1.526
Quantity 25 g
Formula Weight 136.2
Percent Purity 99%
Chemical Name or Material 2-(4-Methylphenyl)ethanol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.