missing translation for 'onlineSavingsMsg'
Learn More

[2-(4-Methylpiperazin-1-yl)phenyl]methanol, 97%, Thermo Scientific™

Product Code. 10619184
Change view
Click to view available options
Quantity:
1 mg
250 mg
10 g
Unit Size:
10g
1g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10619184 1 mg 1g
10567863 250 mg 250mg
10337354 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10619184 Supplier Thermo Scientific Maybridge Supplier No. CC36009DA
 Restricted Item
Estimated Shipment: 12-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 123987-12-2
Molecular Formula C12H19N2O
Molecular Weight (g/mol) 207.30
MDL Number MFCD03407383
InChI Key TWPYBKBPHCMUIS-UHFFFAOYSA-O
Synonym 2-4-methylpiperazin-1-yl phenyl methanol, 2-4-methylpiperazin-1-yl benzyl alcohol, 2-4-methylpiperazino benzyl alcohol, 2-4-methylpiperazin-1yl phenyl methanol, pubchem12999, 2-4-methylpiperazin benzyl alcohol, 2-4-n-methylpiperazinyl benzyl alcohol, 2-4-methyl-1-piperazinyl phenyl methanol, 2-4-methylpiperazinyl phenyl methan-1-ol
PubChem CID 2795567
SMILES C[NH+]1CCN(CC1)C1=CC=CC=C1CO

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 1 mg
IUPAC Name [2-(4-methylpiperazin-1-yl)phenyl]methanol
Formula Weight 206.29
Percent Purity 97%
Chemical Name or Material [2-(4-Methylpiperazin-1-yl)phenyl]methanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.