missing translation for 'onlineSavingsMsg'
Learn More

2-(4-Morpholinyl)ethyl isothiocyanate, 97%, Thermo Scientific Chemicals

Product Code. 11358585
Change view
Click to view available options
Quantity:
2 g
10 g
Unit Size:
10g
2g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11358585 2 g 2g
11368585 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11358585 Supplier Thermo Scientific Alfa Aesar Supplier No. L12046.04
 Restricted Item
Estimated Shipment: 02-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 63224-35-1 | Molecular Formula: C7H12N2OS | Molecular Weight: 172.25 g/mol

2-(4-Morpholinyl)ethyl isothiocyanate is used as pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-(4-Morpholinyl)ethyl isothiocyanate is used as pharmaceutical intermediates.

Solubility
Hydrolyzes in water.

Notes
Store at 4°C. Store under dry inert gas. Protect from humidity and water. Incompatible with moisture, amines, alcohols and oxidizing agents. Protect from heat. Moisture Sensitive.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 63224-35-1
Molecular Formula C7H12N2OS
Molecular Weight (g/mol) 172.25
MDL Number MFCD00041221
InChI Key SDNSCXCXVQHBGH-UHFFFAOYSA-N
Synonym 2-morpholinoethyl isothiocyanate, 4-2-isothiocyanatoethyl morpholine, 2-4-morpholino ethyl isothiocyanate, 2-morpholinoethylisothiocyanate, 4-2-isothiocyanato-ethyl-morpholine, 2-morpholin-4-ylethanisothiocyanate, 2-4-morpholinyl ethyl isothiocyanate, 4-2-isothiocyanatoethyl morpholine #
PubChem CID 143957
IUPAC Name 4-(2-isothiocyanatoethyl)morpholine
SMILES S=C=NCCN1CCOCC1

Specifications

Density 1.11
Boiling Point 162°C to 163°C (10 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.539
Quantity 2 g
UN Number UN2922
Beilstein 1210530
Sensitivity Moisture sensitive
Solubility Information Hydrolyzes in water.
Formula Weight 172.25
Percent Purity 97%
Chemical Name or Material 2-(4-Morpholinyl)ethyl isothiocyanate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.