missing translation for 'onlineSavingsMsg'
Learn More

2-(4-Nitrophenyl)ethanol, 98%

Product Code. 15480788
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15480788 25 g 25g
15460788 1 g 1g
15470788 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15480788 Supplier Thermo Scientific Alfa Aesar Supplier No. H54504.14
 Restricted Item
Estimated Shipment: 21-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 100-27-6 | C8H9NO3 | 167.164 g/mol

It is a phosphate protecting reagent and O6 of deoxyguanosine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is a phosphate protecting reagent and O6 of deoxyguanosine.

Solubility
Slightly soluble in water.

Notes
Incompatible with oxidizing agents. Keep at room temperature.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 100-27-6
Molecular Formula C8H9NO3
Molecular Weight (g/mol) 167.164
MDL Number MFCD00010202
InChI Key IKMXRUOZUUKSON-UHFFFAOYSA-N
Synonym 4-nitrophenethyl alcohol, 2-4-nitrophenyl ethanol, 4-nitrobenzeneethanol, 2-p-nitrophenyl ethanol, benzeneethanol, 4-nitro, p-nitrophenethyl alcohol, 2-4-nitrophenyl ethan-1-ol, phenethyl alcohol, p-nitro, ccris 6079, 4-nitrophenethylalcohol
PubChem CID 7494
IUPAC Name 2-(4-nitrophenyl)ethanol
SMILES C1=CC(=CC=C1CCO)[N+](=O)[O-]

Specifications

Melting Point 59°C to 62°C
Boiling Point 177°C (16 mmHg)
Quantity 25 g
Beilstein 1866148
Solubility Information Slightly soluble in water.
Formula Weight 167.16
Percent Purity 98%
Chemical Name or Material 2-(4-Nitrophenyl)ethanol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.