missing translation for 'onlineSavingsMsg'
Learn More

2,4-Pentanedione, 99%

Product Code. 11454080
Change view
Click to view available options
Quantity:
100 mL
500 mL
2500 mL
Unit Size:
100mL
2500mL
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11454080 100 mL 100mL
11488076 500 mL 500mL
11498076 2500 mL 2500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
Product Code. 11454080 Supplier Thermo Scientific Alfa Aesar Supplier No. A14117.AE
 Restricted Item
Estimated Shipment: 16-07-2026
Add to basket
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

CAS: 123-54-6 | C5H8O2 | 100.12 g/mol

2,4-Pentanedione is used as a precursor to acetylacetonate, a common bidentate and a raw material for the synthesis of heterocyclic compounds. It is used in pharmaceuticals, organic intermediates for solvents, analytical reagents, tungsten and molybdenum in aluminum extraction agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,4-Pentanedione is used as a precursor to acetylacetonate, a common bidentate and a raw material for the synthesis of heterocyclic compounds. It is used in pharmaceuticals, organic intermediates for solvents, analytical reagents, tungsten and molybdenum in aluminum extraction agent.

Solubility
It is readily miscible with water.

Notes
Incompatible with isocyanates, aldehydes, cyanides, peroxides, and anhydrides.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 123-54-6
Molecular Formula C5H8O2
Molecular Weight (g/mol) 100.12
MDL Number MFCD00008787
InChI Key YRKCREAYFQTBPV-UHFFFAOYSA-N
Synonym 2,4-pentanedione, acetylacetone, 2,4-pentadione, diacetylmethane, acetoacetone, acac, 2,4-dioxopentane, pentanedione, pentan-2,4-dione, acetyl acetone
PubChem CID 31261
ChEBI CHEBI:14750
IUPAC Name pentane-2,4-dione
SMILES CC(=O)CC(C)=O

Specifications

Melting Point -23°C
Density 0.973
Boiling Point 140°C to 141°C
Flash Point 34°C (93°F)
Assay Percent Range 99%
Refractive Index 1.452
Quantity 100 mL
UN Number UN2310
Beilstein 741937
Merck Index 14,81
Solubility Information It is readily miscible with water.
Formula Weight 100.12
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 2,4-Pentanedione
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2,4-Pentanedione
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P311-Call a POISON CENTER/doctor/ .
Supplemental information
  • MIXTURE LIST-Contain: 2,4-Pentanedione

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.