missing translation for 'onlineSavingsMsg'
Learn More

2-(4-Penten-1-yl)-1H-isoindole-1,3(2H)-dione, TRC

Product Code. 30179553
Change view
Click to view available options
Quantity:
2.5 g
Unit Size:
2.5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30179553 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30179553 Supplier Toronto Research Chemicals Supplier No. TRCP2275152.5G
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 7736-25-6
Molecular Formula C60 H64 Cl14 Mn N12 O8
Quantity 2.5 g
Synonym 2-(Pent-4-en-1-yl)isoindoline-1,3-dione, N-4-Pentenyl-phthalimide, 5-Phthalimido-1-pentene
InChI Formula InChI=1S/4C15H16Cl3N3O2.2ClH.Mn/c4*1-2-4-20(15(22)21-5-3-19-10-21)6-7-23-14-12(17)8-11(16)9-13(14)18;;;/h4*3,5,8-10H,2,4,6-7H2,1H3;2*1H;/q;;;;;;+2/p-2
SMILES [Cl-][Mn+2]([Cl-])(\[O]=C(\N(CCC)CCOc1c(Cl)cc(Cl)cc1Cl)/n2ccnc2)(\[O]=C(\N(CCC)CCOc3c(Cl)cc(Cl)cc3Cl)/n4ccnc4)(\[O]=C(\N(CCC)CCOc5c(Cl)cc(Cl)cc5Cl)/n6ccnc6)\[O]=C(/N(CCC)CCOc7c(Cl)cc(Cl)cc7Cl)\n8ccnc8
Molecular Weight (g/mol) 1632.506
Formula Weight 1624.999
Chemical Name or Material 2-(4-Penten-1-yl)-1H-isoindole-1,3(2H)-dione
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.