missing translation for 'onlineSavingsMsg'
Learn More

2-(4-pyridyl)-1,3-thiazole-4-carboxylic acid, Thermo Scientific™

Product Code. 10790364
Change view
Click to view available options
Quantity:
250 mg
1 g
10 g
Unit Size:
10g
1g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10790364 250 mg 250mg
10526713 1 g 1g
10605165 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10790364 Supplier Thermo Scientific Maybridge Supplier No. CC34701CB
 Restricted Item
Estimated Shipment: 15-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 21278-86-4
Molecular Formula C9H5N2O2S
Molecular Weight (g/mol) 205.21
MDL Number MFCD00171745
InChI Key COOQMBOJAAZEIR-UHFFFAOYSA-M
Synonym 2-pyridin-4-yl thiazole-4-carboxylic acid, 2-4-pyridyl thiazole-4-carboxylic acid, 2-pyridin-4-yl-1,3-thiazole-4-carboxylic acid, 2-4-pyridyl-1,3-thiazole-4-carboxylic acid, 2-4-pyridinyl-1,3-thiazole-4-carboxylic acid, 2-pyridin-4-yl-thiazole-4-carboxylic acid, 4-thiazolecarboxylicacid, 2-4-pyridinyl, acmc-1cr1v, 4-thiazolecarboxylic acid, 2-4-pyridinyl, 4-carboxy-2-4-pyridyl thiazole
PubChem CID 716091
IUPAC Name 2-pyridin-4-yl-1,3-thiazole-4-carboxylic acid
SMILES [O-]C(=O)C1=CSC(=N1)C1=CC=NC=C1

Specifications

CAS Min % 94.5
CAS Max % 100.0
Packaging Amber glass bottle
Quantity 250 mg
Formula Weight 206.22
Percent Purity 95%
Chemical Name or Material 2-(4-pyridyl)-1, 3-thiazole-4-carboxylic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.