missing translation for 'onlineSavingsMsg'
Learn More

2-(4-Pyridyl)thiazole-4-carboxylic acid, 97%

Product Code. 11484427
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11484427 5 g 5g
11474427 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11484427 Supplier Thermo Scientific Alfa Aesar Supplier No. B20457.06
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 21278-86-4 | C9H5N2O2S | 205.21 g/mol

2-(4-Pyridyl)thiazole-4-carboxylic acid, is used as pharmaceutical intermediate and also used in the field of organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-(4-Pyridyl)thiazole-4-carboxylic acid, is used as pharmaceutical intermediate and also used in the field of organic synthesis.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under normal temperatures and pressures. Keep away from strong oxidizing agents, strong acids, strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 21278-86-4
Molecular Formula C9H5N2O2S
Molecular Weight (g/mol) 205.21
MDL Number MFCD00171745
InChI Key COOQMBOJAAZEIR-UHFFFAOYSA-M
Synonym 2-pyridin-4-yl thiazole-4-carboxylic acid, 2-4-pyridyl thiazole-4-carboxylic acid, 2-pyridin-4-yl-1,3-thiazole-4-carboxylic acid, 2-4-pyridyl-1,3-thiazole-4-carboxylic acid, 2-4-pyridinyl-1,3-thiazole-4-carboxylic acid, 2-pyridin-4-yl-thiazole-4-carboxylic acid, 4-thiazolecarboxylicacid, 2-4-pyridinyl, acmc-1cr1v, 4-thiazolecarboxylic acid, 2-4-pyridinyl, 4-carboxy-2-4-pyridyl thiazole
PubChem CID 716091
SMILES [O-]C(=O)C1=CSC(=N1)C1=CC=NC=C1

Specifications

Melting Point >300°C (decomposition)
Quantity 5 g
Solubility Information Insoluble in water.
IUPAC Name 2-pyridin-4-yl-1,3-thiazole-4-carboxylic acid
Formula Weight 206.22
Percent Purity 97%
Chemical Name or Material 2-(4-Pyridyl)thiazole-4-carboxylic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.