missing translation for 'onlineSavingsMsg'
Learn More

2-((4-(tert-Butoxycarbonyl)-3-fluorophenyl)amino)-2-methylpropanoic Acid, TRC

Product Code. 30335861
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30335861 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30335861 Supplier Toronto Research Chemicals Supplier No. TRCB80984350MG
 Restricted Item
Estimated Shipment: 10-09-2026
Add to basket
Add to basket
This item is not returnable. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Tekniske data

Chemical Name or Material 2-((4-(tert-Butoxycarbonyl)-3-fluorophenyl)amino)-2-methylpropanoic Acid
CAS 2512205-23-9
Molecular Formula C15 H20 F N O4
Quantity 50 mg
InChI Formula InChI=1S/C15H20FNO4/c1-14(2,3)21-12(18)10-7-6-9(8-11(10)16)17-15(4,5)13(19)20/h6-8,17H,1-5H3,(H,19,20)
SMILES CC(C)(C)OC(=O)c1ccc(NC(C)(C)C(=O)O)cc1F
IUPAC Name 2-[3-fluoro-4-[(2-methylpropan-2-yl)oxycarbonyl]anilino]-2-methylpropanoic acid
Molecular Weight (g/mol) 297.322
Formula Weight 297.138
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.