missing translation for 'onlineSavingsMsg'
Learn More

2,4-Thiazolidinedione, 99%

Product Code. 11465593
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11465593 100 g 100g
11455593 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11465593 Supplier Thermo Scientific Alfa Aesar Supplier No. A13496.22
 Restricted Item
Estimated Shipment: 09-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 2295-31-0 | C3H3NO2S | 117.122 g/mol

2,4-Thiazolidinedione may be used in the synthesis of sixteen 5-arylidene-2,4-thiazolidinediones, via ultrasound-promoted aldol condensation reaction. It may be used as starting material for the synthesis of drugs with antihyperglycemic activity.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,4-Thiazolidinedione may be used in the synthesis of sixteen 5-arylidene-2,4-thiazolidinediones, via ultrasound-promoted aldol condensation reaction. It may be used as starting material for the synthesis of drugs with antihyperglycemic activity.

Solubility
Soluble in water (30 mg/ml at 3°C), methanol, DMSO, ethanol, and ethyl ether.

Notes
Keep container tightly closed. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2295-31-0
Molecular Formula C3H3NO2S
Molecular Weight (g/mol) 117.122
MDL Number MFCD00005478
InChI Key ZOBPZXTWZATXDG-UHFFFAOYSA-N
Synonym 2,4-thiazolidinedione, thiazolidinedione, thiazolidine-2,4-dione, 2,4-dioxothiazolidine, 2,4 3h,5h-thiazoledione, usaf ek-5496, thiazolidindione, unii-aa68lxk93c, 2,4 thiazolidinedione, thiazolidinedione-2,4
PubChem CID 5437
ChEBI CHEBI:50992
IUPAC Name 1,3-thiazolidine-2,4-dione
SMILES C1C(=O)NC(=O)S1

Specifications

Melting Point 123°C to 126°C
Boiling Point 178°C to 179°C (19 mmHg)
Quantity 100 g
Beilstein 110700
Solubility Information Soluble in water (30mg/ml at 3°C),methanol,DMSO,ethanol,and ethyl ether.
Formula Weight 117.13
Percent Purity 99%
Chemical Name or Material 2,4-Thiazolidinedione

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.