missing translation for 'onlineSavingsMsg'
Learn More

2,4-Xylenesulfonic Acid, TRC

Product Code. 30455841
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30455841 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30455841 Supplier Toronto Research Chemicals Supplier No. TRCX749510500MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2,4-Xylenesulfonic Acid
CAS 88-61-9
Molecular Formula C8 H10 O3 S . H2 O
Quantity 500 mg
Synonym Benzenesulfonic acid','2,4-dimethyl- (9CI','ACI), 2,4-Dimethylbenzenesulfonic acid (ACI), 2,4-Xylenesulfonic acid (7CI','8CI), 2,4-Dimethylbenzene-1-sulfonic acid, m-Xylene-4-sulfonic acid, m-Xylenesulfonic acid, SXA 60
InChI Formula InChI=1S/C8H10O3S.H2O/c1-6-3-4-8(7(2)5-6)12(9,10)11;/h3-5H,1-2H3,(H,9,10,11);1H2
SMILES O.Cc1ccc(c(C)c1)S(=O)(=O)O
IUPAC Name 2,4-dimethylbenzenesulfonic acid;hydrate
Molecular Weight (g/mol) 204.243
Formula Weight 204.0456
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.