missing translation for 'onlineSavingsMsg'
Learn More

2,5-Bis(2,2,2-trifluoroethoxy)benzoic Acid, TRC

Product Code. 30135434
Change view
Click to view available options
Quantity:
10 g
25 g
100 g
Unit Size:
100g
10g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30135434 100 g 100g
30134930 25 g 25g
30009471 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30135434 Supplier Toronto Research Chemicals Supplier No. TRCB585485100G
 Restricted Item
Estimated Shipment: 17-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 35480-52-5
Molecular Formula C11 H8 F6 O4
Molecular Weight (g/mol) 318.17
Synonym 2,5-Bis(2,2,2-trifluoroethoxy)benzoic Acid, Flecainide Acetate Imp. D (EP)
IUPAC Name 2,5-bis(2,2,2-trifluoroethoxy)benzoic acid
SMILES OC(=O)c1cc(OCC(F)(F)F)ccc1OCC(F)(F)F

Specifications

Quantity 100 g
InChI Formula InChI=1S/C11H8F6O4/c12-10(13,14)4-20-6-1-2-8(7(3-6)9(18)19)21-5-11(15,16)17/h1-3H,4-5H2,(H,18,19)
Formula Weight 318.0327
Chemical Name or Material 2,5-Bis(2,2,2-trifluoroethoxy)benzoic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.