missing translation for 'onlineSavingsMsg'
Learn More

2,5-Bis(5-tert-butylbenzooxazol-2-yl)thiophene, TRC

Product Code. 30331985
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30331985 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30331985 Supplier Toronto Research Chemicals Supplier No. TRCB58532010G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2,5-Bis(5-tert-butyl-benzoxazol-2-yl)thiophene (BBOT)
CAS 7128-64-5
Molecular Formula C26 H26 N2 O2 S
Quantity 10 g
Synonym Benzoxazole, 2,2'-(2,5-thiophenediyl)bis[5-tert-butyl- (7CI,8CI), 2,2'-(2,5-Thiophenediyl)bis[5-(1,1-dimethylethyl)benzoxazole], 2,2'-(2,5-Thiophenediyl)bis(5-tert-butylbenzoxazole), 2,2'-(2,5-Thiophenediyl)bis[5-tert-butylbenzoxazole], 2,5-Bis(5-tert-butyl-2-benzoxazolyl)thiophene, 2,5-Bis(5-tert-butylbenzooxazol-2-yl)thiophene, 2,5-Bis(5'-tert-butyl-2-benzoxazolyl)thiophene, 2,5-Di(5-tert-butylbenzoxazol-2-yl)thiophene, B 0047, BBOT, BBOT 150, Benetex OB, Brightener OB, C.I. 515245, C.I. FBA 184, C.I. Fluorescent Brightener 184, C.I. Fluorescent Brightening Agent 184, Fluorescent Brightener 184, Fluorescent brightener OB, Helios SC, Hitex OB, Keyfluor White PL, MPI Bright 100, NSC 158163, O-B TECH, OB, OB 184, OB-FBA 184, S-FN, SpeedBright OB 184, TBO, Tinopal OB, Uvitex 3BSA, Uvitex OB, Uvitex OB 55, 2,5-Bis(5-tert-butyl-benzoxazol-2-yl)thiophene (BBOT)
InChI Formula InChI=1S/C26H26N2O2S/c1-25(2,3)15-7-9-19-17(13-15)27-23(29-19)21-11-12-22(31-21)24-28-18-14-16(26(4,5)6)8-10-20(18)30-24/h7-14H,1-6H3
SMILES CC(C)(C)c1ccc2oc(nc2c1)c3ccc(s3)c4oc5ccc(cc5n4)C(C)(C)C
IUPAC Name 5-tert-butyl-2-[5-(5-tert-butyl-1,3-benzoxazol-2-yl)thiophen-2-yl]-1,3-benzoxazole
Molecular Weight (g/mol) 430.56
Formula Weight 430.1715
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.