missing translation for 'onlineSavingsMsg'
Learn More

2,5-Bis(tert-butyldioxy)-2,5-dimethylhexane (>90%), TRC

Product Code. 30329449
Change view
Click to view available options
Quantity:
10 mL
Unit Size:
10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30329449 10 mL 10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30329449 Supplier Toronto Research Chemicals Supplier No. TRCB78637010ML

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2,5-Bis(tert-butyldioxy)-2,5-dimethylhexane
CAS 78-63-7
Molecular Formula C16 H34 O4
Quantity 10 mL
Synonym 2,5-Bis(tert-butylperoxy)-2,5-dimethylhexane, Luperox 101-50D, Luperox 101PP10, 2,5-Di(t-butylperoxy)-2,5-dimethylhexane, Varox 50, Varox DBPH, 2,5-Di(tert-butylperoxy)-2,5-dimethylhexane, 2,5-Di-tert-butyl-2,5-dimethylhexyl Peroxide, 2,5-Dimethyl-2,5-bis(1,1-dimethylethylperoxy)hexane, 2,5-Dimethyl-2,5-bis(tert-butyldioxy)hexane, 2,5-Dimethyl-2,5-bis(tert-butylperoxide)hexanel, 2,5-Dimethyl-2,5-bis(tert-butylperoxy)hexane, 2,5-Dimethyl-2,5-di(t-butylperoxy)hexane, 2,5-Dimethyl-2,5-di(tert-butylperoxy)hexane, 2,5-Dimethyl-di(tert-butyl)peroxyhexane, 2,5-Dimethylhexane-2,5-di-tert-butylperoxide, 2,5-Methyl-2,5-di(tert-butylperoxy)hexane, 25B40 AD, AD 40C, APO, APO 10DL, APO 40S, BPDH C 15, C 15 (peroxide), C 4, C 8, C 8 (Vulcanizer), C 8A, CR 05, CT 8, CT 8 (crosslinking agent), Colink 101-
InChI Formula InChI=1S/C16H34O4/c1-13(2,3)17-19-15(7,8)11-12-16(9,10)20-18-14(4,5)6/h11-12H2,1-10H3
SMILES CC(C)(C)OOC(C)(C)CCC(C)(C)OOC(C)(C)C
IUPAC Name 2,5-bis(tert-butylperoxy)-2,5-dimethylhexane
Molecular Weight (g/mol) 290.4388
Formula Weight 290.2457
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.