missing translation for 'onlineSavingsMsg'
Learn More

2,5-Bis(tert-butyldioxy)-2,5-dimethylhexane (>90%), TRC

Product Code. 30346500
Change view
Click to view available options
Quantity:
5 mL
Unit Size:
5mL
Product Code. Quantity unitSize
30346500 5 mL 5mL
1 options
This item is not returnable. View return policy

Product Code. 30346500

Brand: Toronto Research Chemicals TRCB7863705ML

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2,5-Bis(tert-butyldioxy)-2,5-dimethylhexane
CAS 78-63-7
Molecular Formula C16 H34 O4
Quantity 5 mL
Synonym 2,5-Bis(tert-butylperoxy)-2,5-dimethylhexane, Luperox 101-50D, Luperox 101PP10, 2,5-Di(t-butylperoxy)-2,5-dimethylhexane, Varox 50, Varox DBPH, 2,5-Di(tert-butylperoxy)-2,5-dimethylhexane, 2,5-Di-tert-butyl-2,5-dimethylhexyl Peroxide, 2,5-Dimethyl-2,5-bis(1,1-dimethylethylperoxy)hexane, 2,5-Dimethyl-2,5-bis(tert-butyldioxy)hexane, 2,5-Dimethyl-2,5-bis(tert-butylperoxide)hexanel, 2,5-Dimethyl-2,5-bis(tert-butylperoxy)hexane, 2,5-Dimethyl-2,5-di(t-butylperoxy)hexane, 2,5-Dimethyl-2,5-di(tert-butylperoxy)hexane, 2,5-Dimethyl-di(tert-butyl)peroxyhexane, 2,5-Dimethylhexane-2,5-di-tert-butylperoxide, 2,5-Methyl-2,5-di(tert-butylperoxy)hexane, 25B40 AD, AD 40C, APO, APO 10DL, APO 40S, BPDH C 15, C 15 (peroxide), C 4, C 8, C 8 (Vulcanizer), C 8A, CR 05, CT 8, CT 8 (crosslinking agent), Colink 101-
InChI Formula InChI=1S/C16H34O4/c1-13(2,3)17-19-15(7,8)11-12-16(9,10)20-18-14(4,5)6/h11-12H2,1-10H3
SMILES CC(C)(C)OOC(C)(C)CCC(C)(C)OOC(C)(C)C
IUPAC Name 2,5-bis(tert-butylperoxy)-2,5-dimethylhexane
Molecular Weight (g/mol) 290.4388
Formula Weight 290.2457
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.