missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals 2-(5-Bromo-2-pyridylazo)-5-(diethylamino)phenol, 98%

Product Code. 10351151 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10351151 1 g Glass bottle 1g
10742475 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10351151 Supplier Thermo Scientific Chemicals Supplier No. 403220010
 Restricted Item
Estimated Shipment: 28-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 14337-53-2 | C15H17BrN4O | 349.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14337-53-2
Molecular Formula C15H17BrN4O
Molecular Weight (g/mol) 349.23
MDL Number MFCD00006255
InChI Key LYVSIKOGJUDRBK-QGOAFFKASA-N
Synonym 2-5-bromo-2-pyridylazo-5-diethylamino phenol, 2-5-bromopyridin-2-yl diazenyl-5-diethylamino phenol, bromo-padap, 2-5-bromo-2-pyridylazo-5-diethylaminophenol, phenol, 2-5-bromo-2-pyridinyl azo-5-diethylamino, 2-4-diethylamino-2-hydroxyphenylazo-5-bromopyridine, hnvcxdavehoibp-vhebqxmusa-n, 2-5-bromo-2-pyridylazo-5-diethylamino-phenol, 5-diethylamino-2-5-bromo 2-pyridyl diazenyl phenol, e-2-5-bromopyridin-2-yl diazenyl-5-diethylamino phenol
PubChem CID 5911417
IUPAC Name (6E)-6-[(5-bromopyridin-2-yl)hydrazinylidene]-3-(diethylamino)cyclohexa-2,4-dien-1-one
SMILES CCN(CC)C1=CC(=O)\C(C=C1)=N\NC1=CC=C(Br)C=N1

Specifications

CAS Min % 99.0
CAS Max % 100.0
Melting Point 155.0°C to 160.0°C
Color Red
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 349.22
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 2-(5-Bromo-2-pyridylazo)-5-(diethylamino)phenol
Show More Show Less
compliance-icons
Product Identifier
  • 2-(5-Bromo-2-pyridylazo)-5-(diethylamino)phenol
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.