missing translation for 'onlineSavingsMsg'
Learn More

2,5-Di(2,2,2-trifluoroethoxy)benzene-1-carbohydrazide, ≥95%, Thermo Scientific™

Product Code. 10273422
Change view
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10273422 10 g Amber Glass Bottle 10g
10354682 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10273422 Supplier Thermo Scientific Maybridge Supplier No. SPB03377EA
 Restricted Item
Estimated Shipment: 07-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 50778-75-1
Molecular Formula C11H10F6N2O3
Molecular Weight (g/mol) 332.202
MDL Number MFCD00221192
InChI Key WJYCMPVGQTYGSQ-UHFFFAOYSA-N
Synonym 2,5-bis 2,2,2-trifluoroethoxy benzoic acid hydrazide, 2,5-bis 2,2,2-trifluoroethoxy benzohydrazide, 2,5-di 2,2,2-trifluoroethoxy benzene-1-carbohydrazide, maybridge1_008849, 2,5-bis 2,2,2-tris fluoranyl ethoxy benzohydrazide, 2,5-bis 2,2,2-trifluoroethoxy benzenecarbohydrazide, 2,5-bis 2,2,2-trifluoroethoxy benzoic acid hydrazi, 2,5-bis 2,2,2-trifluoroethoxy benzoic acid hydrazide 95+%, 2,5-bis 2,2,2-trifluoroethoxy benzoic acidhydrazide
PubChem CID 3547540
IUPAC Name 2,5-bis(2,2,2-trifluoroethoxy)benzohydrazide
SMILES C1=CC(=C(C=C1OCC(F)(F)F)C(=O)NN)OCC(F)(F)F

Specifications

CAS Min % 90.0
Assay Percent Range 90%+
Packaging Amber Glass Bottle
Quantity 10 g
Formula Weight 332.2
Percent Purity 90%
Chemical Name or Material 2,5-Di(2,2,2-trifluoroethoxy)benzene-1-carbohydrazide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.