missing translation for 'onlineSavingsMsg'
Learn More

2,5-Diaminoanisole Sulfate, TRC

Product Code. 30007622
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30007622 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30007622 Supplier Toronto Research Chemicals Supplier No. TRCD4150001G
 Restricted Item
Estimated Shipment: 11-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 66671-82-7
Molecular Formula C7H10N2O . (H2O4S)
Quantity 1 g
Synonym 2-Methoxy-1,4-benzenediamine Sulfate, 2-Methoxy-p-phenylenediamine Sulfate, Methoxy-p-phenylenediamine Sulfate;
InChI Formula InChI=1S/C7H10N2O.H2O4S/c1-10-7-4-5(8)2-3-6(7)9;1-5(2,3)4/h2-4H,8-9H2,1H3;(H2,1,2,3,4)
SMILES COc1cc(N)ccc1N.OS(=O)(=O)O
Molecular Weight (g/mol) 236.25
Formula Weight 236.0467
Chemical Name or Material 2,5-Diaminoanisole Sulfate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.