missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dibromo-3,4-dinitrothiophene, 95%

Product Code. 11368385
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11368385 1 g 1g
11378385 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11368385 Supplier Thermo Scientific Alfa Aesar Supplier No. L10348.03
 Restricted Item
Estimated Shipment: 21-07-2026
Add to basket
This item is not returnable. View return policy

CAS: 52431-30-8 | C4Br2N2O4S | 331.92 g/mol

Reatant in the synthesis of bromodinitro compound, polymers via stille coupling. 3,4-diaminothiophene was obtained from the reduction of 2,5-dibromo-3,4-dinitrothiophene.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Reatant in the synthesis of bromodinitro compound, polymers via stille coupling. 3,4-diaminothiophene was obtained from the reduction of 2,5-dibromo-3,4-dinitrothiophene.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 52431-30-8
Molecular Formula C4Br2N2O4S
Molecular Weight (g/mol) 331.92
MDL Number MFCD00015537
InChI Key AHGHPBPARMANQD-UHFFFAOYSA-N
Synonym 2,5-dibromo-3,4-dinitro-thiophene, thiophene, 2,5-dibromo-3,4-dinitro, acmc-1apgk, ksc491i5j, 2,5-dibromo-3,4-dinitrothiophene gc
PubChem CID 257024
IUPAC Name 2,5-dibromo-3,4-dinitrothiophene
SMILES [O-][N+](=O)C1=C(Br)SC(Br)=C1[N+]([O-])=O

Specifications

Melting Point 134°C to 137°C
Quantity 1 g
Beilstein 218219
Solubility Information Insoluble in water.
Formula Weight 331.94
Percent Purity 95%
Chemical Name or Material 2,5-Dibromo-3,4-dinitrothiophene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.