missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dibromo-p-xylene, 98+%

Product Code. 11311407
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11311407 25 g 25g
11321407 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11311407 Supplier Thermo Scientific Alfa Aesar Supplier No. L01776.14
 Restricted Item
Estimated Shipment: 17-07-2026
Add to basket
This item is not returnable. View return policy

CAS: 1074-24-4 | C8H8Br2 | 263.96 g/mol

2,5-Dibromo-p-xylene is used as a liquid crystal material. It is also used as an intermediate in agrochemicals and in pharmaceuticals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,5-Dibromo-p-xylene is used as a liquid crystal material. It is also used as an intermediate in agrochemicals and in pharmaceuticals.

Solubility
Soluble in toluene, hot ethanol and hot methanol.Insoluble in water.

Notes
Incompatible with strong oxidizing agents, strong acids and strong alkalis.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1074-24-4
Molecular Formula C8H8Br2
Molecular Weight (g/mol) 263.96
MDL Number MFCD00000091
InChI Key QENIALCDPFDFHX-UHFFFAOYSA-N
Synonym 2,5-dibromo-p-xylene, benzene, 1,4-dibromo-2,5-dimethyl, p-xylene, 2,5-dibromo, 2,5-dibromo-1,4-dimethylbenzene, 2,5-dimethyl-1,4-dibromobenzene, 2,5-dibromo-4-xylene, 2,4-dibromobenzene, p-xylene,5-dibromo, pubchem14829, acmc-1c2tg
PubChem CID 66175
IUPAC Name 1,4-dibromo-2,5-dimethylbenzene
SMILES CC1=CC(Br)=C(C)C=C1Br

Specifications

Melting Point 72°C to 74°C
Quantity 25 g
Odor Odorless
Beilstein 2080433
Boiling Point 261°C
Solubility Information Soluble in toluene,hot ethanol and hot methanol.Insoluble in water.
Formula Weight 263.97
Percent Purity ≥98%
Chemical Name or Material 2,5-Dibromo-p-xylene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.