missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dichloroaniline, 99%

Product Code. 11429073
Change view
Click to view available options
Quantity:
250 g
500 g
2500 g
Unit Size:
2500g
250g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11429073 2500 g 2500g
11414080 250 g 250g
11419073 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11429073 Supplier Thermo Scientific Alfa Aesar Supplier No. A11151.0E
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 95-82-9 | C6H5Cl2N | 162.01 g/mol

2,5-Dichloroaniline was used in a study to develop a fast and sensitive quantitative method for the detection of some aniline derivatives by solid-phase microextraction in gas chromatography-mass spectrometry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,5-Dichloroaniline was used in a study to develop a fast and sensitive quantitative method for the detection of some aniline derivatives by solid-phase microextraction in gas chromatography-mass spectrometry.

Solubility
Soluble in dilute hydrochloric acid.

Notes
Air & Light Sensitive, keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 95-82-9
Molecular Formula C6H5Cl2N
Molecular Weight (g/mol) 162.01
MDL Number MFCD00007667
InChI Key AVYGCQXNNJPXSS-UHFFFAOYSA-N
Synonym 1-amino-2,5-dichlorobenzene, azoic diazo component 3, scarlet 2g base, fast scarlet gg base, 2,5-dichloranilin, benzenamine, 2,5-dichloro, azobase dca, scarlet base gg, fast scarlet 2g, scarlet base ngg
PubChem CID 7262
ChEBI CHEBI:34245
IUPAC Name 2,5-dichloroaniline
SMILES NC1=CC(Cl)=CC=C1Cl

Specifications

Melting Point 48°C to 52°C
Density 1.54
Boiling Point 250°C to 251°C
Flash Point 139°C (282°F)
Quantity 2500 g
UN Number UN3442
Beilstein 1447438
Sensitivity Air and light sensitive
Solubility Information Soluble in dilute hydrochloric acid.
Formula Weight 162.02
Percent Purity 99%
Chemical Name or Material 2,5-Dichloroaniline
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.