missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dichlorobenzonitrile, 98%

Product Code. 11418116
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11418116 5 g 5g
11428116 25 g 25g
11438116 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11418116 Supplier Thermo Scientific Alfa Aesar Supplier No. A14141.06
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

CAS: 21663-61-6 | C7H3Cl2N | 172.008 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Identificatori chimici

CAS 21663-61-6
Molecular Formula C7H3Cl2N
Molecular Weight (g/mol) 172.008
MDL Number MFCD00019741
InChI Key LNGWRTKJZCBXGT-UHFFFAOYSA-N
Synonym benzonitrile, 2,5-dichloro, 2,5-dichloro-benzonitrile, 2,5-dichlorobenzonitrite, 2,5-dichlorobenzenecarbonitrile, pubchem3687, acmc-209fn5, ksc490q4h, 2,5-dichlorobenzonitrile, attercop-chm at133057
PubChem CID 89000
IUPAC Name 2,5-dichlorobenzonitrile
SMILES C1=CC(=C(C=C1Cl)C#N)Cl

Specifica

Melting Point 128°C to 131°C
Quantity 5 g
UN Number UN3439
Beilstein 2045293
Formula Weight 172.01
Percent Purity 98%
Chemical Name or Material 2,5-Dichlorobenzonitrile

RUO – Research Use Only

Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.