missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dichlorothiophene-3-sulfonyl chloride, 97%

Product Code. 11372526
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11372526 5 g 5g
11362526 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11372526 Supplier Thermo Scientific Alfa Aesar Supplier No. L06461.06
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 56946-83-9 | C4HCl3O2S2 | 251.52 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56946-83-9
Molecular Formula C4HCl3O2S2
Molecular Weight (g/mol) 251.52
MDL Number MFCD00051665
InChI Key JJKSHSHZJOWSEC-UHFFFAOYSA-N
Synonym 2,5-dichlorothiophene-3-sulphonyl chloride, 2,5-dichloro-3-thiophenesulfonyl chloride, 3-thiophenesulfonyl chloride, 2,5-dichloro, 2,5-dichlorothiophene-3-sulfonic acid chloride, pubchem7412, 2,5-dichlorothiophene-3-sulfonylchloride, acmc-1avwk, 2,5-dichloro 3-thienyl chlorosulfone, 2,5-dichloro-3-thienylsulfonyl chloride, 2,5-dichlorothien-3-yl-sulfonyl chloride
PubChem CID 2736094
IUPAC Name 2,5-dichlorothiophene-3-sulfonyl chloride
SMILES C1=C(SC(=C1S(=O)(=O)Cl)Cl)Cl

Specifications

Density 1.697
Boiling Point 256°C to 257°C
Flash Point >110°C (230°F)
Refractive Index 1.597
Quantity 5 g
UN Number UN3265
Beilstein 1285315
Sensitivity Moisture sensitive
Formula Weight 251.54
Percent Purity 97%
Chemical Name or Material 2,5-Dichlorothiophene-3-sulfonyl chloride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.