missing translation for 'onlineSavingsMsg'
Learn More

2,5-Difluorobenzoic acid, TRC

Product Code. 30374474
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30374474 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30374474 Supplier Toronto Research Chemicals Supplier No. TRCD4511835G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2,5-Difluorobenzoic Acid
CAS 2991-28-8
Molecular Formula C7H4F2O2
Quantity 5 g
Synonym NSC 190310
InChI Formula InChI=1S/C7H4F2O2/c8-4-1-2-6(9)5(3-4)7(10)11/h1-3H,(H,10,11)
SMILES FC1=CC(C(O)=O)=C(F)C=C1
IUPAC Name 2,5-difluorobenzoic acid
Molecular Weight (g/mol) 158.1
Formula Weight 158.0179
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.