missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ 2,5-Dihydro-2,5-dimethoxyfuran, cis + trans, 99%

Product Code. 15443277
Change view
Click to view available options
Quantity:
50 g
Unit Size:
50g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15443277 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15443277 Supplier Thermo Scientific™ Supplier No. B25028.18
 Restricted Item
Estimated Shipment: 07-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 332-77-4 | C6H10O3 | 130.14 g/mol

TRUSTED_SUSTAINABILITY

Specifications

CAS 332-77-4
Melting Point <-40°
Density 1.072
Boiling Point 160-162°
Flash Point 47°(116°F)
Molecular Formula C6H10O3
MDL Number MFCD00003220
Quantity 50 g
UN Number UN1993
Synonym 2,5-Dimethoxy-2,5-dihydrofuran;
InChI Key WXFWXFIWDGJRSC-UHFFFAOYNA-N
SMILES COC1OC(OC)C=C1
IUPAC Name 2,5-dimethoxy-2,5-dihydrofuran
Molecular Weight (g/mol) 130.14
Formula Weight 130.14
Percent Purity 99%
Physical Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.