missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dihydro-2,5-dimethoxyfuran, cis + trans, 99%

Product Code. 11444070
Change view
Click to view available options
Quantity:
100 g
250 g
500 g
1000 g
Unit Size:
1000g
100g
250g
500g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11444070 100 g 100g
11495987 250 g 250g
11454070 500 g 500g
11485987 1000 g 1000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11444070 Supplier Thermo Scientific Alfa Aesar Supplier No. B25028.22
 Restricted Item
Estimated Shipment: 29-07-2026
Add to basket
This item is not returnable. View return policy

CAS: 332-77-4 | C6H10O3 | 130.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 332-77-4
Molecular Formula C6H10O3
Molecular Weight (g/mol) 130.14
MDL Number MFCD00003220
InChI Key WXFWXFIWDGJRSC-UHFFFAOYNA-N
Synonym 2,5-dihydro-2,5-dimethoxyfuran, furan, 2,5-dihydro-2,5-dimethoxy, 2,5-dihydro-2,5-dimethoxyfuran, cis+trans, 2,5-dihydro-2,5-dimethoxyfuran, mixture of cis and trans, pubchem6933, ksc490q3d, furan,5-dihydro-2,5-dimethoxy, 2,5-dihydro-2,5-dimethoxyfuran,c&t, 2,5-dimethoxy-2,5-dihydrofuran,c&t, 2,5-dihydro-2,5-dimethoxyfuran, cis + trans
PubChem CID 78974
IUPAC Name 2,5-dimethoxy-2,5-dihydrofuran
SMILES COC1OC(OC)C=C1

Specifications

Melting Point <-40°C
Density 1.072
Boiling Point 160°C to 162°C
Flash Point 47°C (116°F)
Refractive Index 1.435
Quantity 100 g
UN Number UN3271
Beilstein 113225
Formula Weight 130.14
Percent Purity 99%
Chemical Name or Material 2,5-Dihydro-2,5-dimethoxyfuran, cis + trans
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.