missing translation for 'onlineSavingsMsg'
Learn More

2,5-Diiodo-1-methylimidazole, 98%, Thermo Scientific Chemicals

Product Code. 15438077
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15438077 1 g 1g
15448077 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15438077 Supplier Thermo Scientific Alfa Aesar Supplier No. H51212.03
 Restricted Item
Estimated Shipment: 16-09-2026
Add to basket
This item is not returnable. View return policy

It is employed as intermediate for pharmaceutical.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed as intermediate for pharmaceutical.

Notes
Light Sensitive, store in dark. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 86026-81-5
Molecular Formula C4H4I2N2
Molecular Weight (g/mol) 333.899
MDL Number MFCD02179540
InChI Key SZYKHAFVWJNSQU-UHFFFAOYSA-N
PubChem CID 335838
IUPAC Name 2,5-diiodo-1-methylimidazole
SMILES CN1C(=CN=C1I)I

Specifications

Quantity 1 g
Sensitivity Light sensitive
Synonym 2,5-diiodo-1-methyl-1h-imidazole, pubchem8993, acmc-1bke8, 1-methyl-2,5-diiodoimidazole, 1-methyl-2,5-diiodo-1h-imidazole, 1h-imidazole,2,5-diiodo-1-methyl, 2,5-bis iodanyl-1-methyl-imidazole, 1h-imidazole, 2,5-diiodo-1-methyl, 2,5-diiodo-1-methylimidazole
Formula Weight 333.9
Percent Purity 98%
Chemical Name or Material 2,5-Diiodo-1-methylimidazole

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.