missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dimercapto-1,3,4-thiadiazole, 97% (dry wt.), water <3%

Product Code. 11490433
Change view
Click to view available options
Quantity:
50 g
250 g
Unit Size:
250g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11490433 50 g 50g
11400443 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11490433 Supplier Thermo Scientific Alfa Aesar Supplier No. A12615.18
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1072-71-5 | C2H2N2S3 | 150.23 g/mol

2,5-Dimercapto-1,3,4-thiadiazole is used in synthesizing polymers and heavy metal and basic salts. It acts as an additive in lubricating oils, greases and in electrode compositions. It also acts as an intermediate for pharmaceuticals and dyes, as a chelating agent in the analysis of metals. It also serves as an anticorrosion and antirust agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,5-Dimercapto-1,3,4-thiadiazole is used in synthesizing polymers and heavy metal and basic salts. It acts as an additive in lubricating oils, greases and in electrode compositions. It also acts as an intermediate for pharmaceuticals and dyes, as a chelating agent in the analysis of metals. It also serves as an anticorrosion and antirust agent.

Solubility
Soluble in water.

Notes
Incompatible with strong oxidizing agents and strong acids.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1072-71-5
Molecular Formula C2H2N2S3
Molecular Weight (g/mol) 150.23
MDL Number MFCD00003103
InChI Key BIGYLAKFCGVRAN-UHFFFAOYSA-N
Synonym 1,3,4-thiadiazole-2,5-dithiol, 2,5-dimercapto-1,3,4-thiadiazole, bismuthiol i, dimercaptothiadiazole, bismuththiol, 2,5-dimercaptothiadiazole, usaf a-8354, bismuthiol, py 61h, bismuththiol i
PubChem CID 2723630
IUPAC Name 1,3,4-thiadiazolidine-2,5-dithione
SMILES S=C1NNC(=S)S1

Specifications

Melting Point ∼162°C (decomposition)
Quantity 50 g
Beilstein 113444
Solubility Information Soluble in water.
Formula Weight 150.24
Percent Purity 98%
Chemical Name or Material 2,5-Dimercapto-1,3,4-thiadiazole

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.