missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dimercapto-1,3,4-thiadiazole, 98%

Product Code. 10653363 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Plastic bottle
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10653363 5 g Glass bottle 5g
10520571 25 g Plastic bottle 25g
10060810 100 g Plastic bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10653363 Supplier Thermo Scientific Acros Supplier No. 115310050
 Restricted Item
Estimated Shipment: 28-09-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1072-71-5 | C2H2N2S3 | 150.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1072-71-5
Molecular Formula C2H2N2S3
Molecular Weight (g/mol) 150.23
MDL Number MFCD00003103
InChI Key BIGYLAKFCGVRAN-UHFFFAOYSA-N
Synonym 1,3,4-thiadiazole-2,5-dithiol, 2,5-dimercapto-1,3,4-thiadiazole, bismuthiol i, dimercaptothiadiazole, bismuththiol, 2,5-dimercaptothiadiazole, usaf a-8354, bismuthiol, py 61h, bismuththiol i
PubChem CID 2723630
IUPAC Name 1,3,4-thiadiazolidine-2,5-dithione
SMILES S=C1NNC(=S)S1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 161°C to 165°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Quantity 5 g
Beilstein 27, 677
Solubility Information Solubility in water: 20g/L water (20°C)
Formula Weight 150.23
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 2,5-Dimercapto-1,3,4-thiadiazole, 98%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.