missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,5-Dimethoxytetrahydrofuran, 99%, mixture of cis- and trans isomers
Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
500g
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Spécification
Spécification
| Melting Point | -45°C |
| CAS Min % | 98.5 |
| Color | Colorless to Yellow |
| CAS Max % | 100.0 |
| Density | 1.0210g/mL |
| Boiling Point | 145°C to 148°C |
| Quantity | 100 g |
| Infrared Spectrum | Authentic |
| Packaging | Glass bottle |
| Physical Form | Liquid |
| Afficher plus |
Sécurité et traitement
Identifiant produit
- 2,5-Dimethoxytetrahydrofuran, mixture of cis- and trans isomers
- Warning
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 3
- H319-Causes serious eye irritation.
- H226-Flammable liquid and vapour.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu