missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dimethoxytetrahydrofuran, 99%, mixture of cis- and trans isomers

Product Code. 10773651 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
500g
Product Code. Quantity Packaging unitSize
10773651 500 g Glass bottle 500g
10294330 100 g Glass bottle 100g
2 options
This item is not returnable. View return policy

Product Code. 10773651

Brand: Thermo Scientific Acros 115675000

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 696-59-3 | C6H12O3 | 132.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 696-59-3
Molecular Formula C6H12O3
Molecular Weight (g/mol) 132.16
MDL Number MFCD00005359
InChI Key GFISDBXSWQMOND-UHFFFAOYSA-N
Synonym 2,5-dimethoxytetrahydrofuran, tetrahydro-2,5-dimethoxyfuran, furan, tetrahydro-2,5-dimethoxy, 2,5-dimethoxytetrahydrofurane, 2,5-dimethoxy-tetrahydrofuran, 2,5-dimethoxytetrahydrofuran, mixture of cis-and trans isomers, dimethoxytetrahydrofuran van, protectol dmt, furan,5-dimethoxy, dmthf
PubChem CID 79098
IUPAC Name 2,5-dimethoxyoxolane
SMILES COC1CCC(O1)OC

Specifications

Melting Point -45°C
CAS Min % 98.5
Color Colorless to Yellow
CAS Max % 100.0
Density 1.0210g/mL
Boiling Point 145°C to 148°C
Quantity 500 g
Infrared Spectrum Authentic
Packaging Glass bottle
Physical Form Liquid
Refractive Index 1.4160 to 1.419
Assay Percent Range 99%
Fieser 02,144
Solubility Information Solubility in water: 350g/L (20°C). Other solubilities: miscible with alcohols and most common organic,solvents
Formula Weight 132.16
Percent Purity 99%
Chemical Name or Material 2, 5-Dimethoxytetrahydrofuran, 99%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2,5-Dimethoxytetrahydrofuran, mixture of cis- and trans isomers
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.