missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2,5-Dimethyl-1-(4-nitrophenyl)pyrrole, 97%

Product Code. 11456217
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11456217 5 g 5g
11446217 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11456217 Supplier Alfa Aesar™ Supplier No. B25145.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 5044-22-4
Molecular Formula C12H12N2O2
Molecular Weight (g/mol) 216.24
MDL Number MFCD00085038
InChI Key JXJRCFOGAIXRCU-UHFFFAOYSA-N
Synonym 2,5-dimethyl-1-4-nitrophenyl-1h-pyrrole, 2,5-dimethyl-1-4-nitrophenyl pyrrole, 1-4-nitrophenyl-2,5-dimethylpyrrole, maybridge1_007286, acmc-1auq2, 2,5-dimethyl-1-p-nitrophenyl pyrrole, n-4'-nitrophenyl-2,5-dimethylpyrrole, 1-4-nitrophenyl-2,5-dimethyl-1h-pyrrole
PubChem CID 459085
IUPAC Name 2,5-dimethyl-1-(4-nitrophenyl)pyrrole
SMILES CC1=CC=C(N1C2=CC=C(C=C2)[N+](=O)[O-])C

Specifications

Melting Point 145°C to 148°C
Quantity 5 g
Formula Weight 216.24
Percent Purity 97%
Chemical Name or Material 2,5-Dimethyl-1-(4-nitrophenyl)pyrrole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.