missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dimethyl-3-hexyne-2,5-diol, (+/-) + meso, 98%

Product Code. 11305339
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11305339 500 g 500g
11395329 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11305339 Supplier Thermo Scientific Alfa Aesar Supplier No. B20889.36
 Restricted Item
Estimated Shipment: 30-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 142-30-3 | C8H14O2 | 142.198 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 142-30-3
Molecular Formula C8H14O2
Molecular Weight (g/mol) 142.198
MDL Number MFCD00004468
InChI Key IHJUECRFYCQBMW-UHFFFAOYSA-N
Synonym 2,5-dimethyl-3-hexyne-2,5-diol, dimethylhexynediol, kemitracin-50, tetramethylbutynediol, acetylenepinacol, 3-hexyne-2,5-diol, 2,5-dimethyl, olfine y, tetramethyl-2-butynediol, unii-22rr53u71w, 2,5-dimethylhexyne-2,5-diol
PubChem CID 8883
IUPAC Name 2,5-dimethylhex-3-yne-2,5-diol
SMILES CC(C)(C#CC(C)(C)O)O

Specifications

Melting Point 90°C to 94°C
Density 0.949
Boiling Point 205°C
Flash Point 102°C (215°F)
Quantity 500 g
Beilstein 969835
Formula Weight 142.2
Percent Purity 98%
Chemical Name or Material 2,5-Dimethyl-3-hexyne-2,5-diol, (+/-) + meso

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.