missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dimethyl-3-thiophenesulfonyl chloride, 97%, Thermo Scientific™

Product Code. 10198494
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10198494 10 g 10g
10466852 1 g 1g
10412484 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10198494 Supplier Thermo Scientific Maybridge Supplier No. MO00053EA
 Restricted Item
Estimated Shipment: 25-09-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 97272-04-3
Molecular Formula C6H7ClO2S2
Molecular Weight (g/mol) 210.69
MDL Number MFCD03086201
InChI Key CMTPCYKEUFDVAU-UHFFFAOYSA-N
PubChem CID 2779650
IUPAC Name 2,5-dimethylthiophene-3-sulfonyl chloride
SMILES CC1=CC(=C(C)S1)S(Cl)(=O)=O

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Quantity 10 g
Synonym 2,5-dimethyl-3-thiophenesulfonyl chloride, 2,5-dimethylthiophene-3-sulphonyl chloride, 3-thiophenesulfonylchloride, 2,5-dimethyl, buttpark 9650-20, chembrdg-bb 4009594, 2,5-dimethyl 3-thienyl chlorosulfone, 2,5-dimethyl-3-thienyl sulfonyl chloride, 2, 5-dimethylthiophene-3-sulphonyl chloride, 3-thiophenesulfonylchloride,2,5-dimethyl-9ci
Formula Weight 210.7
Percent Purity 97%
Chemical Name or Material 2, 5-Dimethyl-3-thiophenesulfonyl chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.