missing translation for 'onlineSavingsMsg'
Learn More

2',5'-Dimethylacetophenone, 97%

Product Code. 11460283
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11460283 25 g 25g
11450283 5 g 5g
11470283 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11460283 Supplier Thermo Scientific Alfa Aesar Supplier No. A11432.14
 Restricted Item
Estimated Shipment: 02-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 2142-73-6 | C10H12O | 148.205 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2142-73-6
Molecular Formula C10H12O
Molecular Weight (g/mol) 148.205
MDL Number MFCD00014988
InChI Key AWKBVLVKQQRRFQ-UHFFFAOYSA-N
Synonym 2',5'-dimethylacetophenone, 2,5-dimethylacetophenone, 1-2,5-dimethylphenyl ethanone, 1-2,5-dimethylphenyl ethan-1-one, ethanone, 1-2,5-dimethylphenyl, acetophenone, 2',5'-dimethyl, 1-acetyl-2,5-dimethylbenzene, 2-acetyl-p-xylene, acmc-1cliy, 2',5'dimethylacetophenone
PubChem CID 75061
IUPAC Name 1-(2,5-dimethylphenyl)ethanone
SMILES CC1=CC(=C(C=C1)C)C(=O)C

Specifications

Density 1
Boiling Point 230°C to 231°C
Flash Point 95°C (203°F)
Odor Characteristic
Refractive Index 1.529
Quantity 25 g
Beilstein 2042139
Formula Weight 148.21
Percent Purity ≥98%
Chemical Name or Material 2',5'-Dimethylacetophenone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.