missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dimethylaniline, 98+%

Product Code. 11440573
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11440573 500 g 500g
15433057 25 g 25g
11430573 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11440573 Supplier Thermo Scientific Alfa Aesar Supplier No. A12682.36
 Restricted Item
Estimated Shipment: 22-07-2026
Add to basket
This item is not returnable. View return policy

CAS: 95-78-3 | C8H11N | 121.18 g/mol

2,5-Dimethylaniline is widely used as raw materials to produce imaging chemicals like organic dyes and pigments. It is also used in the production of antioxidants, pharmaceutical, agricultural, rubber chemicals and other target organic molecules. 2,5-Dimethylaniline was also used in the synthesis of nanocomposite of multi-walled carbon nanotubes embedded in poly(2,5-dimethylaniline). 2,5-Dimethylaniline was used to study the effect of metabolites formed from 2,5-xylidine by fungi on laccase activity. The Fourier transform infrared (FTIR) and Raman spectra of 2,5-dimethylaniline was studied5. It induced transcription of lccIV.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,5-Dimethylaniline is widely used as raw materials to produce imaging chemicals like organic dyes and pigments. It is also used in the production of antioxidants, pharmaceutical, agricultural, rubber chemicals and other target organic molecules. 2,5-Dimethylaniline was also used in the synthesis of nanocomposite of multi-walled carbon nanotubes embedded in poly(2,5-dimethylaniline). 2,5-Dimethylaniline was used to study the effect of metabolites formed from 2,5-xylidine by fungi on laccase activity. The Fourier transform infrared (FTIR) and Raman spectra of 2,5-dimethylaniline was studied5. It induced transcription of lccIV.

Solubility
Soluble in water <0.1 g/100 mL at 18°C.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 95-78-3
Molecular Formula C8H11N
Molecular Weight (g/mol) 121.18
MDL Number MFCD00007743
InChI Key VOWZNBNDMFLQGM-UHFFFAOYSA-N
Synonym 2,5-xylidine, p-xylidine, 2,5-dimethylphenylamine, 2-amino-1,4-xylene, 2,5-dimethylbenzenamine, benzenamine, 2,5-dimethyl, 5-methyl-o-toluidine, 6-methyl-m-toluidine, 1-amino-2,5-dimethylbenzene, p-dimethylaniline
PubChem CID 7259
ChEBI CHEBI:518305
IUPAC Name 2,5-dimethylaniline
SMILES CC1=CC=C(C)C(N)=C1

Specifications

Melting Point 15°C
Density 0.973
Quantity 500 g
Odor Characteristic
Refractive Index 1.559
UN Number UN1711
Beilstein 2205178
Merck Index 14,10084
Boiling Point 218°C
Solubility Information Soluble in water <0.1g/100 mL at 18°C.
Formula Weight 121.18
Percent Purity ≥98%
Chemical Name or Material 2,5-Dimethylaniline
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.