missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dimethylaniline, 99%

Product Code. 10665192 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
500g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10665192 500 g Glass bottle 500g
10346690 5 g Glass bottle 5g
10356690 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10665192 Supplier Thermo Scientific Acros Supplier No. 146695000
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 95-78-3 | C8H11N | 121.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 95-78-3
Molecular Formula C8H11N
Molecular Weight (g/mol) 121.18
MDL Number MFCD00007743
InChI Key VOWZNBNDMFLQGM-UHFFFAOYSA-N
Synonym 2,5-xylidine, p-xylidine, 2,5-dimethylphenylamine, 2-amino-1,4-xylene, 2,5-dimethylbenzenamine, benzenamine, 2,5-dimethyl, 5-methyl-o-toluidine, 6-methyl-m-toluidine, 1-amino-2,5-dimethylbenzene, p-dimethylaniline
PubChem CID 7259
ChEBI CHEBI:518305
IUPAC Name 2,5-dimethylaniline
SMILES CC1=CC=C(C)C(N)=C1

Specifications

Melting Point 11.5°C
CAS Min % 98.5
CAS Max % 100.0
Density 0.9700g/mL
Quantity 500 g
Infrared Spectrum Authentic
Packaging Glass bottle
Physical Form Liquid
Refractive Index 1.5582 to 1.5602
Assay Percent Range 98.5% min. (GC)
Specific Gravity 0.97
Linear Formula (CH3)2C6H3NH2
Beilstein 12, 1135
Merck Index 15, 10282
Boiling Point 218°C
Solubility Information Solubility in water: slightly soluble. Other solubilities: soluble in alcohol,ether,carbon tetrachloride
Formula Weight 121.18
Percent Purity 99%
Chemical Name or Material 2, 5-Dimethylaniline, 99%
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 2,5-Dimethylaniline
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • Acute toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H330-Fatal if inhaled.
  • H373-May cause damage to organs.
  • H411-Toxic to aquatic life with long lasting effects.
  • H311-Toxic in contact with skin.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.