missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dimethylfuran-3-carbonyl chloride, 97%, Thermo Scientific™

Product Code. 10772473
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10772473 1 g 1g
10448704 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10772473 Supplier Thermo Scientific Maybridge Supplier No. CC01102DA
 Restricted Item
Estimated Shipment: 12-10-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 50990-93-7
Molecular Formula C7H7ClO2
Molecular Weight (g/mol) 158.58
MDL Number MFCD00221073
InChI Key RWORXZHVOUPMMM-UHFFFAOYSA-N
Synonym 2,5-dimethyl-3-furoyl chloride, 2,5-dimethyl-furan-3-carbonyl chloride, 2,5-dimethyl-3-furancarbonyl chloride, 2,5-dimethylfuran-3-carbonylchloride, pubchem6936, acmc-209krc, 2,5-dimethyl-3-furoylchloride, 2,5 dimethyl-furan-3-carbonyl chloride, 2,5-dimethyl furan-3-carbonyl chloride, 3-furancarbonylchloride, 2,5-dimethyl
PubChem CID 2734804
IUPAC Name 2,5-dimethylfuran-3-carbonyl chloride
SMILES CC1=CC(C(Cl)=O)=C(C)O1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Density 1.19g/mL
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 1 g
Formula Weight 158.58
Percent Purity 97%
Chemical Name or Material 2, 5-Dimethylfuran-3-carbonyl chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.