missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dimethylphenol, 99+%

Product Code. 10010910 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
500g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10010910 100 g Glass bottle 100g
10407110 5 g Glass bottle 5g
10020910 500 g Glass bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10010910 Supplier Thermo Scientific Acros Supplier No. 116541000
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 95-87-4 | C8H10O | 122.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 95-87-4
Molecular Formula C8H10O
Molecular Weight (g/mol) 122.17
MDL Number MFCD00002237
InChI Key NKTOLZVEWDHZMU-UHFFFAOYSA-N
Synonym 2,5-xylenol, p-xylenol, phenol, 2,5-dimethyl, 3,6-dimethylphenol, 6-methyl-m-cresol, 3,6-xylenol, 2,5-dimethyl phenol, 2,5-dmp, 1,2,5-xylenol, 1-hydroxy-2,5-dimethylbenzene
PubChem CID 7267
IUPAC Name 2,5-dimethylphenol
SMILES CC1=CC=C(C)C(O)=C1

Specifications

Melting Point 75°C to 77°C
CAS Min % 99.0
Color White to Cream
CAS Max % 100.0
Density 0.9700g/mL
Quantity 100 g
Infrared Spectrum Authentic
Packaging Glass bottle
Physical Form Crystals
Assay Percent Range 99% min. (GC)
Specific Gravity 0.97
Linear Formula (CH3)2C6H3OH
Viscosity 1.7 mPa.s (75°C)
Beilstein 06, 494
Merck Index 15, 10279
Boiling Point 212°C
Solubility Information Solubility in water: 1% (60°C). Other solubilities: soluble in naoh solution,freely soluble in alcohol,chloroform,ether and,benzene
Formula Weight 122.17
Percent Purity 99+%
Chemical Name or Material 2, 5-Dimethylphenol, 99+%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.