missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,5-Dimethylphenol, TRC
Click to view available options
Quantity:
10 g
Unit Size:
10g
Specifications
Specifications
| Chemical Name or Material | 2,5-Dimethylphenol |
| CAS | 95-87-4 |
| Molecular Formula | C8 H10 O |
| Quantity | 10 g |
| Synonym | 2,5-Dimethylphenol, Gemfibrozil Imp. A (EP), p-Xylenol, 2,5-Xylenol (8CI), 1,2,5-Xylenol, 1-Hydroxy-2,5-dimethylbenzene, 2-Hydroxy-p-xylene, 3,6-Dimethylphenol, NSC 2599 |
| InChI Formula | InChI=1S/C8H10O/c1-6-3-4-7(2)8(9)5-6/h3-5,9H,1-2H3 |
| SMILES | Cc1ccc(C)c(O)c1 |
| IUPAC Name | 2,5-dimethylphenol |
| Molecular Weight (g/mol) | 122.16 |
| Formula Weight | 122.0732 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction