missing translation for 'onlineSavingsMsg'
Learn More

(2,5-Dimethylphenyl)boronic Acid, TRC

Product Code. 30374620
Click to view available options
Quantity:
25 g
Unit Size:
25g
This item is not returnable. View return policy

Product Code. 30374620

Brand: Toronto Research Chemicals TRCD47324525G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (2,5-Dimethylphenyl)boronic Acid
CAS 85199-06-0
Molecular Formula C8H11BO2
Quantity 25 g
Synonym B-(2,5-Dimethylphenyl)-Boronic Acid, 2,5-Xyleneboronic Acid, (2,5-Dimethylphenyl)-boronic Acid, (2,5-Dimethylphenyl)boronic Acid, p-Xyleneboronic Acid
InChI Formula InChI=1S/C8H11BO2/c1-6-3-4-7(2)8(5-6)9(10)11/h3-5,10-11H,1-2H3
SMILES Cc1ccc(C)c(c1)B(O)O
IUPAC Name (2,5-dimethylphenyl)boronic acid
Molecular Weight (g/mol) 149.98
Formula Weight 150.0852
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.