missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(2,5-Dimethylphenyl)boronic Acid, TRC
Click to view available options
Quantity:
1 g
Unit Size:
1g
Specifications
Specifications
| Chemical Name or Material | (2,5-Dimethylphenyl)boronic Acid |
| CAS | 85199-06-0 |
| Molecular Formula | C8H11BO2 |
| Quantity | 1 g |
| Synonym | B-(2,5-Dimethylphenyl)-Boronic Acid, 2,5-Xyleneboronic Acid, (2,5-Dimethylphenyl)-boronic Acid, (2,5-Dimethylphenyl)boronic Acid, p-Xyleneboronic Acid |
| InChI Formula | InChI=1S/C8H11BO2/c1-6-3-4-7(2)8(5-6)9(10)11/h3-5,10-11H,1-2H3 |
| SMILES | Cc1ccc(C)c(c1)B(O)O |
| IUPAC Name | (2,5-dimethylphenyl)boronic acid |
| Molecular Weight (g/mol) | 149.98 |
| Formula Weight | 150.0852 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction