missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dimethylphenyl isocyanate, 97%

Product Code. 11322847
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11322847 1 g 1g
11332847 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11322847 Supplier Thermo Scientific Alfa Aesar Supplier No. L15999.03
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 40397-98-6 | C9H9NO | 147.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 40397-98-6
Molecular Formula C9H9NO
Molecular Weight (g/mol) 147.18
MDL Number MFCD00013854
InChI Key SOXVXJQIQVOCAY-UHFFFAOYSA-N
Synonym 2,5-dimethylphenyl isocyanate, 2,5-dimethylphenylisocyanate, benzene, 2-isocyanato-1,4-dimethyl, 2,5-dimethyl phenylisocyanate, 2,5-dimethylbenzenisocyanate, acmc-1art9, 1-isocyanato-2,5-dimethyl-benzene
PubChem CID 98605
ChEBI CHEBI:60097
IUPAC Name 2-isocyanato-1,4-dimethylbenzene
SMILES CC1=CC=C(C)C(=C1)N=C=O

Specifications

Density 1.043
Boiling Point 210°C
Quantity 1 g
Refractive Index 1.5315
UN Number UN2206
Beilstein 1100841
Formula Weight 147.18
Percent Purity 97%
Chemical Name or Material 2,5-Dimethylphenyl isocyanate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.