missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dimethylpyrazine, 99%

Product Code. 10367640 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10367640 5 g Glass bottle 5g
10695042 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10367640 Supplier Thermo Scientific Acros Supplier No. 174520050
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 123-32-0 | C6H8N2 | 108.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 123-32-0
Molecular Formula C6H8N2
Molecular Weight (g/mol) 108.14
MDL Number MFCD00006147
InChI Key LCZUOKDVTBMCMX-UHFFFAOYSA-N
Synonym 2,5-dimethyl pyrazine, pyrazine, 2,5-dimethyl, 2,5-dimethylpiazine, 2,5-dimethyl-1,4-diazine, 2,5-dimethylparadiazine, 2,5-dimethyl-pyrazine, unii-v99y0muy1q, fema no. 3272, ccris 2929, natural
PubChem CID 31252
IUPAC Name 2,5-dimethylpyrazine
SMILES CC1=CN=C(C=N1)C

Specifications

CAS Min % 98.5
CAS Max % 100.0
Color Colorless to Yellow
Density 0.9900g/mL
Boiling Point 155°C
Flash Point 63°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Refractive Index 1.4987 to 1.5007
Quantity 5 g
Beilstein 23, 96
Specific Gravity 0.99
Solubility Information Solubility in water: soluble. Other solubilities: soluble in organic solvents,miscible with ethanol,ethylether,acetone
Formula Weight 108.14
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 2, 5-Dimethylpyrazine, 99%
Show More Show Less
compliance-icons
Product Identifier
  • 2,5-Dimethylpyrazine
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P264-Wash face, hands and any thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.