missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dimethylpyrazine, 99%

Product Code. 11449483
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11449483 100 g 100g
11429483 5 g 5g
11439483 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11449483 Supplier Thermo Scientific Alfa Aesar Supplier No. A12352.22
 Restricted Item
Estimated Shipment: 18-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 123-32-0 | C6H8N2 | 108.144 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 123-32-0
Molecular Formula C6H8N2
Molecular Weight (g/mol) 108.144
MDL Number MFCD00006147
InChI Key LCZUOKDVTBMCMX-UHFFFAOYSA-N
Synonym 2,5-dimethyl pyrazine, pyrazine, 2,5-dimethyl, 2,5-dimethylpiazine, 2,5-dimethyl-1,4-diazine, 2,5-dimethylparadiazine, 2,5-dimethyl-pyrazine, unii-v99y0muy1q, fema no. 3272, ccris 2929, natural
PubChem CID 31252
IUPAC Name 2,5-dimethylpyrazine
SMILES CC1=CN=C(C=N1)C

Specifications

Melting Point 15°C
Density 0.99
Boiling Point 154°C to 155°C
Flash Point 63°C (145°F)
Odor Pleasant
Refractive Index 1.5 00
Quantity 100 g
Beilstein 107052
Formula Weight 108.14
Percent Purity 99%
Chemical Name or Material 2,5-Dimethylpyrazine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.