missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dithiobiurea, 97%

Product Code. 11472648
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11472648 100 g 100g
11462648 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11472648 Supplier Thermo Scientific Alfa Aesar Supplier No. A16667.22
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 142-46-1 | C2H6N4S2 | 150.22 g/mol

2,5-Dithiobiurea is used in radiopharmaceutical imaging, inhibiting enzyme function, kidney function study and to treat toxic metal poisoning. It is also used in co-crystals development and in the field of nonlinear optics to generate new coherent wavelengths

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,5-Dithiobiurea is used in radiopharmaceutical imaging, inhibiting enzyme function, kidney function study and to treat toxic metal poisoning. It is also used in co-crystals development and in the field of nonlinear optics to generate new coherent wavelengths

Solubility
Soluble in hot water, pyridine, triethylamine and dimethylformamide. Insoluble in cold water.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 142-46-1
Molecular Formula C2H6N4S2
Molecular Weight (g/mol) 150.22
MDL Number MFCD00014471
InChI Key KCOYHFNCTWXETP-UHFFFAOYSA-N
Synonym 2,5-dithiobiurea, dithiobiurea, bithiourea, dithiourea, 1,2-hydrazinedicarbothioamide, bis thiourea, hydrazodicarbothioamide, 2,5-dithiodiurea, hydrazine-1,2-bis carbothioamide, dithiocarbamoylhydrazine
PubChem CID 2724564
IUPAC Name (carbamothioylamino)thiourea
SMILES NC(=S)NNC(N)=S

Specifications

Melting Point ∼212°C (decomposition)
Quantity 100 g
Beilstein 1723178
Solubility Information Soluble in hot water,pyridine,triethylamine and dimethylformamide. Insoluble in cold water.
Formula Weight 150.22
Percent Purity 97%
Chemical Name or Material 2,5-Dithiobiurea

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.