missing translation for 'onlineSavingsMsg'
Learn More

2-[(5-Fluoresceinyl)aminocarbonyl]ethyl Methanethiosulfonate, TRC

Product Code. 30138104
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30138104 25 mg 25mg
17982245 5 mg 5mg
17913515 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30138104 Supplier Toronto Research Chemicals Supplier No. TRCF51000025MG
 Restricted Item
Estimated Shipment: 15-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 351330-42-2
Molecular Formula C24 H19 N O8 S2
Molecular Weight (g/mol) 513.54
IUPAC Name N-(3',6'-dihydroxy-3-oxospiro[2-benzofuran-1,9'-xanthene]-5-yl)-3-methylsulfonylsulfanylpropanamide
SMILES CS(=O)(=O)SCCC(=O)Nc1ccc2c(c1)C(=O)OC23c4ccc(O)cc4Oc5cc(O)ccc35

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C24H19NO8S2/c1-35(30,31)34-9-8-22(28)25-13-2-5-17-16(10-13)23(29)33-24(17)18-6-3-14(26)11-20(18)32-21-12-15(27)4-7-19(21)24/h2-7,10-12,26-27H,8-9H2,1H3,(H,25,28)
Formula Weight 513.0552
Chemical Name or Material 2-[(5-Fluoresceinyl)aminocarbonyl]ethyl Methanethiosulfonate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.