missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,5-Furandicarboxylic Acid-13C6, TRC
Specifications
Specifications
| Chemical Name or Material | 2,5-Furandicarboxylic Acid-13C6 |
| Molecular Formula | 13C6H4O5 |
| Quantity | 5 mg |
| Synonym | Dehydromucic Acid-13C6, Furane-α,α'-dicarboxylic Acid-13C6, NSC 40740-13C6 |
| InChI Formula | InChI=1S/C6H4O5/c7-5(8)3-1-2-4(11-3)6(9)10/h1-2H,(H,7,8)(H,9,10)/i1+1,2+1,3+1,4+1,5+1,6+1 |
| SMILES | O=[13C](O)[13C]1=[13C]([H])[13C]([H])=[13C]([13C](O)=O)O1 |
| IUPAC Name | furan-2,5-dicarboxylic-2,3,4,5-13C4 acid-2,5-13C2 |
| Molecular Weight (g/mol) | 162.05 |
| Formula Weight | 162.03 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?