missing translation for 'onlineSavingsMsg'
Learn More

2,5-Furandicarboxylic Acid, TRC

Product Code. 30399437
Change view
Click to view available options
Quantity:
1 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30399437 1 g
30399596 25 g
30397662 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30399437 Supplier Toronto Research Chemicals Supplier No. TRCF8637501G
 Restricted Item
Estimated Shipment: 21-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3238-40-2
Molecular Formula C6 H4 O5
Molecular Weight (g/mol) 156.09
Synonym 2,5-Furandicarboxylic acid
IUPAC Name furan-2,5-dicarboxylic acid
SMILES OC(=O)c1oc(cc1)C(=O)O

Specifications

Quantity 1 g
InChI Formula InChI=1S/C6H4O5/c7-5(8)3-1-2-4(11-3)6(9)10/h1-2H,(H,7,8)(H,9,10)
Formula Weight 156.0059
Chemical Name or Material 2,5-Furandicarboxylic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.